siRNA Details
| Gene Symbol: Gm13031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045911.1 | 1 | 36 | |||
| GenBank mRNA | AK040418.1 | - | 1 | 37 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB635979.1 | - | 1 | 37 | Chr. 4 - 140947987 - 140957902 [-] on Build GRCm38 | |

Gm13031
Genomic Map