siRNA Details
| Gene Symbol: Gm13031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045911.1 | 4 | 572 | |||
| GenBank mRNA | AK040418.1 | - | 4 | 573 | ||
| BE651383.1 | - | 3 | 212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13031| Gene Symbol: Gm13031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045911.1 | 4 | 572 | |||
| GenBank mRNA | AK040418.1 | - | 4 | 573 | ||
| BE651383.1 | - | 3 | 212 | |||