siRNA Details
| Gene Symbol: Gnas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045792.1 | - | 1 | 1944 | ||
| AK082220.1 | - | 1 | 2446 | |||
| AK085136.1 | - | 1 | 427 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL593857.10 | - | 189680 | Chr. - - [-] on Build | ||

Gnas
Genomic Map