siRNA Details
| Gene Symbol: Gm33489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146979.1 | - | 3 | 635 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY760728.1 | - | 3 | 611 | Chr. 13 - 44010592 - 44018500 [-] on Build GRCm38 | |

Gm33489
Genomic Map