siRNA Details
| Gene Symbol: 2810036E18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012862.1 | - | 1 | 161 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711973.1 | - | 1 | 161 | Chr. 2 - 130273772 - 130274269 [-] on Build GRCm38 | |

2810036E18Rik
Genomic Map