siRNA Details
Gene Symbol: 2810036E18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012862.1 | - | 1 | 415 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711973.1 | - | 1 | 415 | Chr. 2 - 130273772 - 130274269 [-] on Build GRCm38 |
