siRNA Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015814.1 | - | 935 | |||
| Gene Symbol: Gm26555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029615.1 | - | 6 | 1068 | ||

Kif27
Genomic Map