siRNA Details
| Gene Symbol: Phf21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK157510.1 | - | 1 | 1545 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY570764.1 | - | 1 | 223 | Chr. 2 - 92211887 - 92212356 [+] on Build GRCm38 | |

Phf21a
Genomic Map