siRNA Details
| Gene Symbol: Poglut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076379.1 | - | 1302 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Poglut1| Gene Symbol: Poglut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076379.1 | - | 1302 | |||