siRNA Details
| Gene Symbol: Catsperd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014981.1 | - | 4 | 632 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714503.1 | - | 4 | 633 | Chr. 17 - 56628143 - 56634147 [+] on Build GRCm38 | |

Catsperd
Genomic Map