siRNA Details
| Gene Symbol: Skap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK162116.1 | - | 3 | 611 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ052412.1 | - | 3 | 611 | Chr. 11 - 96483780 - 96492269 [+] on Build GRCm38 | |

Skap1
Genomic Map