siRNA Details
| Gene Symbol: 4930477N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015581.1 | - | 3 | 542 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA464769.1 | - | 5 | 627 | Chr. 10 - 119320234 - 119331845 [+] on Build GRCm38 | |

4930477N07Rik
Genomic Map