siRNA Details
| Gene Symbol: 4930477J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015580.1 | - | 1 | 535 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715130.1 | - | 1 | 535 | Chr. 18 - 36320906 - 36321492 [-] on Build GRCm38 | |

4930477J04Rik
Genomic Map