siRNA Details
| Gene Symbol: 4930478M13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015589.1 | - | 421 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715139.1 | - | 2 | 418 | Chr. 15 - 100105234 - 100108159 [-] on Build GRCm38 | |

4930478M13Rik
Genomic Map