siRNA Details
| Gene Symbol: 4930478M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015588.1 | - | 1 | 456 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715138.1 | - | 1 | 456 | Chr. 5 - 31451513 - 31452331 [-] on Build GRCm38 | |

4930478M09Rik
Genomic Map