siRNA Details
| Gene Symbol: Rest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011349.1 | - | 1 | 1664 | ||
| BC023433.1 | - | 1 | 377 | |||
| BC051648.1 | - | 1 | 713 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rest| Gene Symbol: Rest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011349.1 | - | 1 | 1664 | ||
| BC023433.1 | - | 1 | 377 | |||
| BC051648.1 | - | 1 | 713 | |||