siRNA Details
| Gene Symbol: 4930563H03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA178695.1 | - | 1 | 213 | ||
| AK019780.1 | - | 1 | 133 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715814.1 | - | 1 | 133 | Chr. 15 - 79074706 - 79075048 [-] on Build GRCm38 | |

4930563H03Rik
Genomic Map