siRNA Details
| Gene Symbol: Gm28590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031624.1 | - | 4 | 393 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB621240.1 | - | 4 | 393 | Chr. 1 - 122205145 - 122250480 [+] on Build GRCm38 | |

Gm28590
Genomic Map