siRNA Details
| Gene Symbol: Rbm46os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040381.1 | 4 | 381 | |||
| NR_040382.1 | 4 | 372 | ||||
| GenBank mRNA | AK019784.1 | - | 3 | 332 | ||
| AK161442.1 | - | 4 | 382 | |||
| CJ047138.1 | - | 3 | 215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm46os| Gene Symbol: Rbm46os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040381.1 | 4 | 381 | |||
| NR_040382.1 | 4 | 372 | ||||
| GenBank mRNA | AK019784.1 | - | 3 | 332 | ||
| AK161442.1 | - | 4 | 382 | |||
| CJ047138.1 | - | 3 | 215 | |||