siRNA Details
| Gene Symbol: Rbm46os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040381.1 | 4 | 666 | |||
| NR_040382.1 | 4 | 657 | ||||
| GenBank mRNA | AK019784.1 | - | 3 | 617 | ||
| AK161442.1 | - | 4 | 667 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715831.1 | - | 3 | 615 | Chr. 3 - 82878685 - 82941099 [+] on Build GRCm38 | |

Rbm46os
Genomic Map