siRNA Details
| Gene Symbol: 4930487H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040601.1 | 1 | 160 | |||
| GenBank mRNA | AK015638.1 | - | 1 | 161 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930487H11Rik| Gene Symbol: 4930487H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040601.1 | 1 | 160 | |||
| GenBank mRNA | AK015638.1 | - | 1 | 161 | ||