siRNA Details
Gene Symbol: 4930486I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045734.1 | 3 | 439 | |||
GenBank mRNA | AK015632.1 | - | 3 | 440 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930486I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045734.1 | 3 | 439 | |||
GenBank mRNA | AK015632.1 | - | 3 | 440 |