siRNA Details
| Gene Symbol: Clcn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017285.1 | - | 1 | 323 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK790760.1 | - | 1 | 80 | Chr. X - 7154118 - 7154854 [-] on Build GRCm38 | |

Clcn5
Genomic Map