siRNA Details
| Gene Symbol: Gm19554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043332.1 | - | 2 | 571 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB640799.1 | - | 2 | 571 | Chr. 12 - 105899841 - 105900715 [+] on Build GRCm38 | |

Gm19554
Genomic Map