siRNA Details
| Gene Symbol: Gm19412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140218.1 | - | 1 | 57 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ060377.1 | - | 1 | 57 | Chr. 17 - 33919400 - 33920485 [-] on Build GRCm38 | |

Gm19412
Genomic Map