siRNA Details
| Gene Symbol: 4930566F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016234.1 | - | 1 | 122 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715846.1 | - | 1 | 122 | Chr. 5 - 31487299 - 31495179 [-] on Build GRCm38 | |

4930566F21Rik
Genomic Map