siRNA Details
| Gene Symbol: Trim54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016235.1 | - | 3 | 817 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim54| Gene Symbol: Trim54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016235.1 | - | 3 | 817 | ||