siRNA Details
| Gene Symbol: Trim54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016235.1 | - | 3 | 637 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715847.1 | - | 3 | 635 | Chr. 5 - 31135929 - 31137344 [-] on Build GRCm38 | |

Trim54
Genomic Map