siRNA Details
| Gene Symbol: Gm34184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035016.1 | - | 1 | 46 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB627249.1 | - | 1 | 46 | Chr. 10 - 84038567 - 84100697 [-] on Build GRCm38 | |

Gm34184
Genomic Map