siRNA Details
| Gene Symbol: Ppp1r18os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035015.1 | - | 1 | 1904 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB100689.2 | - | 1 | 649 | Chr. 17 - 35859653 - 35860304 [-] on Build GRCm38 | |

Ppp1r18os
Genomic Map