siRNA Details
| Gene Symbol: Ppp1r18os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035015.1 | - | 1 | 798 | ||
| Gene Symbol: Wdr18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032251.1 | - | 2 | 1253 | ||

Ppp1r18os
Genomic Map