siRNA Details
Gene Symbol: 5033423K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017188.1 | - | 1 | 207 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716918.1 | - | 1 | 207 | Chr. 5 - 33141233 - 33142110 [-] on Build GRCm38 |
