siRNA Details
| Gene Symbol: 6530406A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018324.1 | - | 1 | 1227 | ||
| AK087211.1 | - | 1 | 2721 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB534254.2 | - | 1 | 511 | Chr. 12 - 112955547 - 112956192 [-] on Build GRCm38 | |

6530406A20Rik
Genomic Map