siRNA Details
Gene Symbol: 6530406A20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA739088.1 | - | 1 | 392 | ||
AK018324.1 | - | 1 | 1124 | |||
AK087211.1 | - | 1 | 2618 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6530406A20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA739088.1 | - | 1 | 392 | ||
AK018324.1 | - | 1 | 1124 | |||
AK087211.1 | - | 1 | 2618 |