siRNA Details
| Gene Symbol: 6530406A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018324.1 | - | 1 | 182 | ||
| AK087211.1 | - | 1 | 1674 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6530406A20Rik| Gene Symbol: 6530406A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018324.1 | - | 1 | 182 | ||
| AK087211.1 | - | 1 | 1674 | |||