siRNA Details
| Gene Symbol: Zfp191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017180.1 | - | 1 | 64 | ||
| AK088914.1 | - | 1 | 3402 | |||
| BC044900.1 | - | 1 | 2259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp191| Gene Symbol: Zfp191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017180.1 | - | 1 | 64 | ||
| AK088914.1 | - | 1 | 3402 | |||
| BC044900.1 | - | 1 | 2259 | |||