siRNA Details
| Gene Symbol: Gm19589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037971.1 | 6 | 2214 | |||
| GenBank mRNA | AK030268.1 | - | 6 | 2214 | ||
| AK136671.1 | - | 2 | 1306 | |||
| BB818892.1 | - | 1 | 28 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19589| Gene Symbol: Gm19589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037971.1 | 6 | 2214 | |||
| GenBank mRNA | AK030268.1 | - | 6 | 2214 | ||
| AK136671.1 | - | 2 | 1306 | |||
| BB818892.1 | - | 1 | 28 | |||