siRNA Details
Gene Symbol: 2610507I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037964.1 | 3 | 2177 | |||
GenBank mRNA | AK012065.1 | - | 1 | 1073 | ||
AK162965.1 | - | 3 | 2132 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2610507I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037964.1 | 3 | 2177 | |||
GenBank mRNA | AK012065.1 | - | 1 | 1073 | ||
AK162965.1 | - | 3 | 2132 |