siRNA Details
| Gene Symbol: 2610507I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037964.1 | 3 | 2339 | |||
| GenBank mRNA | AK012065.1 | - | 1 | 1235 | ||
| AK162965.1 | - | 3 | 2294 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ312656.1 | - | 1 | 138 | Chr. 11 - 59197917 - 59198497 [-] on Build GRCm38 | |

2610507I01Rik
Genomic Map