siRNA Details
| Gene Symbol: 2610507I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037964.1 | 3 | 2211 | |||
| GenBank mRNA | AK012065.1 | - | 1 | 1107 | ||
| AK162965.1 | - | 3 | 2166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610507I01Rik| Gene Symbol: 2610507I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037964.1 | 3 | 2211 | |||
| GenBank mRNA | AK012065.1 | - | 1 | 1107 | ||
| AK162965.1 | - | 3 | 2166 | |||