siRNA Details
| Gene Symbol: 2810452K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013332.1 | - | 1 | 798 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ243472.1 | - | 1 | 453 | Chr. 4 - 129624255 - 129624723 [-] on Build GRCm38 | |

2810452K05Rik
Genomic Map