siRNA Details
| Gene Symbol: 2810403D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007586.1 | - | 1 | 82 | ||
| AK012972.1 | - | 1 | 61 | |||
| AK155344.1 | - | 1 | 76 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810403D21Rik| Gene Symbol: 2810403D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007586.1 | - | 1 | 82 | ||
| AK012972.1 | - | 1 | 61 | |||
| AK155344.1 | - | 1 | 76 | |||