siRNA Details
| Gene Symbol: 9530001D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020532.1 | - | 1 | 154 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721963.1 | - | 1 | 154 | Chr. 13 - 54741952 - 54742336 [+] on Build GRCm38 | |

9530001D17Rik
Genomic Map