siRNA Details
| Gene Symbol: Zfp346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084710.1 | - | 1 | 2072 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB478760.2 | - | 1 | 568 | Chr. 13 - 55134504 - 55135170 [+] on Build GRCm38 | |

Zfp346
Genomic Map