siRNA Details
| Gene Symbol: 1700025N23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040523.1 | 4 | 357 | |||
| GenBank mRNA | AK006361.1 | - | 4 | 357 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706428.1 | - | 369 | Chr. - - [-] on Build | ||

1700025N23Rik
Genomic Map