siRNA Details
| Gene Symbol: 1700025N23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040523.1 | 4 | 792 | |||
| GenBank mRNA | AK006361.1 | - | 4 | 792 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700025N23Rik| Gene Symbol: 1700025N23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040523.1 | 4 | 792 | |||
| GenBank mRNA | AK006361.1 | - | 4 | 792 | ||