siRNA Details
| Gene Symbol: 1700025N21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006360.1 | - | 2 | 649 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706427.1 | - | 2 | 649 | Chr. 10 - 83139150 - 83142038 [-] on Build GRCm38 | |

1700025N21Rik
Genomic Map