siRNA Details
| Gene Symbol: Gm30529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045892.1 | - | 2 | 838 | ||
| Gene Symbol: Trim39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053591.1 | - | 1 | 41 | ||

Gm30529
Genomic Map