siRNA Details
| Gene Symbol: Gm30529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045892.1 | - | 1 | 49 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB643625.1 | - | 1 | 49 | Chr. 5 - 112649367 - 112651371 [+] on Build GRCm38 | |

Gm30529
Genomic Map