siRNA Details
| Gene Symbol: Sntg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080155.1 | - | 5 | 1501 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB221905.2 | - | 5 | 461 | Chr. 1 - 8950572 - 9298236 [-] on Build GRCm38 | |

Sntg1
Genomic Map